Order Entry
Puerto Rico
ContactUsLinkComponent
Human Recombinant PTN (Animal free) (from E. coli)
Human Recombinant PTN (Animal free) (from E. coli)
Certificates
undefined
Human Recombinant PTN (Animal free) (from E. coli)

Specifications

  • Conjugation:
    Unconjugated
  • Protein Function:
    Cytokine
  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Storage Conditions:
    −20 °C
  • Endotoxin Content:
    Endotoxin LAL (EU/µg) ≤0.1
  • Biological Activity:
    Bioactivity: No biological activity data is available at this time
  • Gene ID:
    P21246
  • Reconstitution Instructions:
    Sterile water at 0.1 mg/ml
  • Endotoxin-free:
    N
  • Carrier-Free:
    Y
  • Protease-free:
    N
  • Animal-Free:
    Y
  • Protein Synonyms:
    NEGF1|HARP|HBBM|HBGF-8
  • UniProtKB:
    P21246
  • Protein/Peptide Name:
    PTN (AF)
  • Purity:
    ≥95%
  • Molecular Weight:
    15.4 kDa
  • Sequence:
    MGKKEKPEKK VKKSDCGEWQ WSVCVPTSGD CGLGTREGTR TGAECKQTMK TQRCKIPCNW KKQFGAECKY QFQAWGECDL NTALKTRTGS LKRALHNAEC QKTVTISKPC GKLTKPKPQA ESKKKKKEGK KQEKMLD
  • Endotoxin Level:
    Low
  • Formulation:
    10 mM sodium phosphate, pH 7.5
  • Nuclease-free:
    N
  • Grade:
    RUO
  • Shipping Temperature:
    RT

Specifications

Related Information