Human Recombinant B-NGF (from E. coli)
Supplier: FUJIFILM IRVINE SCIENTIFIC, INC
Certificates
About this item
Nerve growth factor beta (β-NGF) is a neurotrophic factor that is important for the development and maintenance of sensory and sympathetic neurons. β-NGF signals through the low affinity nerve growth factor receptor (LNGFR) and the tropomyosin receptor kinase A (TrkA) to activate PI3K, Ras, and PLC signaling pathways. β-NGF is also involved in the growth, differentiation, and survival of B lymphocytes. Human, mouse, and rat β-NGF proteins are cross-reactive.
- High quality
- Low endotoxin
- Affordable
Specifications
- Conjugation:Unconjugated
- Protein Function:Cytokine
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Human
- Storage Conditions:–20 °C
- Endotoxin Content:Endotoxin LAL (EU/µg) ≤0.1
- Biological Activity:Bioactivity Assay: TF-1 cell proliferation, ED₅₀ ≤5 ng/ml, Bioactivity ≥2.0×10⁵ units/mg
- Gene ID:P01138
- Reconstitution Instructions:Sterile water at 0.1 mg/ml
- Endotoxin-free:N
- Carrier-Free:Y
- Protease-free:N
- Animal-Free:Y
- Protein Synonyms:beta-NGF
- Protein/Peptide Name:B-NGF (AF)
- Purity:≥95%
- Molecular Weight:13.6 kDa
- Sequence:MSSSHPIFHR GEFSVCDSVS VWVGDKTTAT DIKGKEVMVL GEVNINNSVF KQYFFETKCR DPNPVDSGCR GIDSKHWNSY CTTTHTFVKA LTMDGKQAAW RFIRIDTACV CVLSRKAVRR A
- Endotoxin Level:Low
- Formulation:0.1% Trifluoroacetic acid (TFA)
- Nuclease-free:N
- Grade:RUO
- Shipping Temperature:RT
- Tested Applications:Bioassay
Specifications
Related Information
Literature reference - PubMed ID 25149537
Additional Product Details
Shenandoah_Biotechnology_Product_Catalog