Human Recombinant RANK LIGAND (Animal free) (from E. coli)
Supplier: FUJIFILM IRVINE SCIENTIFIC, INC
Certificates
About this item
Stromal cell-derived factor-1 alpha (SDF-1 α), also called CXCL12a, is one of two SDF-1 splice variants made by a wide variety of cells upon stimulation by inflammatory cytokines such as TNF, IL-1, and LPS. SDF-1α signals through the G protein-coupled receptor CXCR4 to recruit activated leukocytes. Human and mouse SDF-1 α share 99% sequence identity.
- High quality
- Low endotoxin
- Affordable
Shenandoah Biotechnology is a manufacturer of research grade recombinant proteins typically used as cytokines, growth factors and chemokines in cell based assays and cell culture. Each production lot is tested for QA and the expected values for each product can be viewed under the product specifications tab.
Caution: For research use only.
Specifications
- Conjugation:Unconjugated
- Protein Function:Cytokine
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Human
- Storage Conditions:−20 °C
- Endotoxin Content:Endotoxin LAL (EU/µg) ≤1
- Biological Activity:Bioactivity assay: Ability to induce bioluminescence in engineered RANKL bioassay cells , ED50 ≤50 ng/ml, bioactivity ≥2.0×10⁴ units/mg
- Gene ID:O14788
- Reconstitution Instructions:Sterile water at 0.1 mg/ml
- Endotoxin-free:N
- Carrier-Free:Y
- Protease-free:N
- Animal-Free:Y
- Protein Synonyms:OPGL|TNFSF11|ODF|TRANCE
- UniProtKB:O14788
- Protein/Peptide Name:RANK LIGAND (AF)
- Purity:≥95%
- Molecular Weight:19.7 kDa
- Sequence:EKAMVDGSWL DLAKRSKLEA QPFAHLTINA TDIPSGSHKV SLSSWYHDRG WAKISNMTFS NGKLIVNQDG FYYLYANICF RHHETSGDLA TEYLQLMVYV TKTSIKIPSS HTLMKGGSTK YWSGNSEFHF YSINVGGFFK LRSGEEISIE VSNPSLLDPD QDATYFGAFK VRDID
- Endotoxin Level:Low
- Formulation:10 mM sodium phosphate, pH 7.5
- Nuclease-free:N
- Grade:RUO
- Shipping Temperature:RT
- Tested Applications:Bioassay
Specifications
Related Information
Literature reference - PubMed ID 21191633
Additional Product Details
Shenandoah_Biotechnology_Product_Catalog