Human Recombinant IL-3 (from E. coli)
Supplier: Fujifilm Irvine Scientific
Certificates
About this item
Interleukin 3 (IL-3) is a cytokine that is produced by activated T cells and mast cells. IL-3 induces the differentiation of hematopoietic stem cells into myeloid precursor cells, such as erythrocyte, megakaryocyte, granulocyte, monocyte, and dendritic cells. IL-3 also functions in the nervous system and is important during the B-1 cell regulation of chronic inflammatory diseases.
- High quality
- Low endotoxin
- Affordable
Specifications
- Conjugation:Unconjugated
- Protein Function:Cytokine
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Human
- Storage Conditions:–20 °C
- Endotoxin Content:Endotoxin LAL (EU/µg) ≤0.1
- Biological Activity:Bioactivity Assay: TF-1 cell proliferation, ED₅₀ ≤2 ng/ml, Bioactivity ≥5.0×10⁵ units/mg
- Gene ID:P08700
- Reconstitution Instructions:Sterile water at 0.1 mg/ml
- Endotoxin-free:N
- Carrier-Free:Y
- Protease-free:N
- Animal-Free:Y
- Protein Synonyms:MCGF|P-cell stimulating factor|HCGF|Multi-CSF
- Protein/Peptide Name:IL-3 (AF)
- Purity:≥97%
- Molecular Weight:15.2 kDa
- Sequence:MAPMTQTTPL KTSWVNCSNM IDEIITHLKQ PPLPLLDFNN LNGEDQDILM ENNLRRPNLE AFNRAVKSLQ NASAIESILK NLLPCLPLAT AAPTRHPIHI KDGDWNEFRR KLTFYLKTLE NAQAQQTTLS LAIF
- Endotoxin Level:Low
- Formulation:10 mM Sodium phosphate, 150 mM Sodium chloride, pH 7.5
- Nuclease-free:N
- Grade:RUO
- Shipping Temperature:RT
- Tested Applications:Bioassay
Specifications
Related Information
Literature reference - PubMed ID 24461581
Additional Product Details
Shenandoah_Biotechnology_Product_Catalog
Product Literature
Frequently Bought Together