Order Entry
Puerto Rico
ContactUsLinkComponent
ActiveHOOK™ Bovine Serum Albumin (BSA) (active)
ActiveHOOK™ Bovine Serum Albumin (BSA) (active)
  82022-448
 :  
ActiveHOOK™ Bovine Serum Albumin (BSA) (active)
  82022-448
 :  786-093
 :  

 

  • Source:
    Serum
  • Species:
    Bovine
  • Size:
    2 mg
  • Protein Synonyms:
    BSA|Fraction V|Cohn Fraction V
  • Protein/Peptide Name:
    Bovine Serum Albumin (active)
  • Cat. No.:
    82022-448
  • Supplier no.:
    786-093

 

 

OneQuant ActiveHOOK™ BSA is a pre-activated form of immunological grade BSA (MW 67kD) that has been activated by sulfoSMCC and is ready for direct coupling with peptides through the cysteine residues. Immunological grade BSA is a high purity grade bovine serum albumin (BSA) prepared for immunotechnology research applications such as a carrier protein for antibody production. ActiveHOOK™ BSA is supplied lyophilized in OneQuant™ format; 2mg/vial.