Order Entry
Puerto Rico
ContactUsLinkComponent
OneQuant™ KLH
OneQuant™ KLH
Catalog # 82022-444
Supplier:  G-Biosciences
CAS Number:  
OneQuant™ KLH
Catalog # 82022-444
Supplier:  G-Biosciences
Supplier Number:  786-091
CAS Number:  

Specifications

  • Size:
    2 mg
  • Protein/Peptide Name:
    KLH
  • Grade:
    Immunological Grade
  • Cat. No.:
    82022-444
  • Supplier no.:
    786-091

Specifications

About this item