Specifications
- Conjugation:Biotin
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Size:0.5 mg
- Storage Conditions:Upon receipt store at –20 °C
- Protein/Peptide Name:Protein L
- Molecular Weight:~35,800 (Apparent MW by SDS-PAGE: 36,000)
- Shipping Temperature:ambient
- Cat. No.:PI29997
- Supplier no.:29997
Specifications
About this item
Pierce™ Biotinylated Recombinant Protein L can be used to probe and detect mouse and human antibodies containing kappa light chains in Western blotting, ELISA, IHC and other immunoassay protocols.
- Contains four immunoglobulin binding domains per protein
- Binds members of all classes of Ig that contain kappa light chains
- Binds single chain variable fragments (scFv) and Fab fragments that contain kappa light chains
Protein L is an immunoglobulin-binding protein that was originally derived from the bacteria Peptostreptococcus magnus, but is now produced recombinantly. Protein L has the unique ability to bind through kappa light chain interactions without interfering with an antibody's antigen-binding site. This gives Protein L the ability to bind a wider range of Ig classes and subclasses than other antibody-binding proteins. Protein L will bind to all classes of Ig (IgG, IgM, IgA, IgE and IgD). Protein L will also bind single chain variable fragments (scFv) and Fab fragments. Protein L binds kappa I, III, and IV in human and kappa I on mouse.
Form: salt-free powder
A280 of 0.1% solution:
0.80
Isoelectric point (pI): 4.8