Test strips, peroxide , Reflectoquant®, MilliporeSigma
Catalog # EM1.16731.0001
Supplier: MilliporeSigma
CAS Number:
Specifications
- Description:Test with Strips
- Cat. No.:EM1.16731.0001
- Range:100-1000 mg/L H₂O₂
- Supplier no.:1.16731.0001
- No. of tests:50 strips
Specifications
About this item
The test is suited for the determination of inorganic peroxides in aqueous solutions
- Fast analysis with reliable results
- Barcode calibration
- Low analysis costs
- Environmentally friendly
Furthermore the test can be used in disinfecting and rinsing solutions (e.g. in food technology, laundries) and in swimming-pool water.
Test strips for the reflectometers RQflex® 10, RQflex® 10 plus and RQflex® 20.