About this item
TSKgel ODS-140HTP columns provide high resolution and short analyses time at moderate pressures, enabling high throughput separations
- Polylayer bonding chemistry of the particle size of these columns results in highly efficient and durable columns
- Used with either UPLC® (up to 9000 psi) or conventional HPLC systems
- Excellent peak shape for basic compounds
Ideal for use in high throughput applications including drug discovery, pharmacokinetics and peptide digest separations.