Spectra Antibody Labeling Kit, Azure Biosystems
Supplier: Azure Biosystems
Certificates
About this item
Make your own fluorescently conjugated antibodies!
- Make your own fluorescently conjugated antibodies
- Two labeling reactions for 50 mg of antibody
- Covalently label primary or secondary antibodies
Our labeling kit includes everything you need for covalently labeling primary or secondary antibodies with a fluorescent dye. Choose between visible and near-IR conugates.
Ordering information: Kit includes enough for two labeling reactions for 50 µg of antibody.