GravityFlo® Portable Eyewash, SE-4400, Speakman®
Catalog # 10068-226
Supplier: Speakman
Specifications
- Description:Gravityflo® portable eyewash
- Environmentally Preferable:
- Cat. No.:10068-226
- Supplier no.:SE-4400
Specifications
About this item
The SE-4400 portable eyewash allows to locate an eyewash anywhere that it is needed, even where potable water is not available.
- Features a 34.06 L (9 gal.) tank, providing continuous flow
- Capability to be shelf, table, or wall mounted
- Pull down activation with hands free operation
- Plastic fill cap with integral tamper prevention and molded plastic sprays
Certifications: Meets ANSI/ISEA Z358.1.