AimStep® Cotinine Test, Germaine® Laboratories
Catalog # 10024-940
Supplier: Germaine Laboratories
Specifications
- Description:AimStep® Cotinine Test
- Cat. No.:10024-940
- Supplier no.:40010
Specifications
About this item
AimStep® Cotinine is a rapid qualitative test for the detection of cotinine in urine
Results are available in five minutes. Each test includes an internal procedural control.
The test is designed to detect cotinine at a concentration of 200ng/mL in urine. Each test is individually packed in a foil pouch.