All-in-One Drugs of Abuse Key Test Cups, Germaine® Laboratories
Supplier: Germaine Laboratories
Certificates
About this item
SafeCup™ II Key Cups are a series of All-in-One (collection/testing/transport) qualitative tests for the detection of drugs of abuse in urine
Tests include 25 test cups, security seals, gloves, and instructions.
These tests adhere to SAMSHA cutoff levels. Each cup is leak-proof and equipped with a temperature strip. Tests do not activate until the key is turned. Cups feature a flat side for easy photocopying for permanent record.
Certifications: The SafeCup™ II MultiDrug 5 test is CLIA waived.
Frequently Bought Together