Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Azure Fluorescent Blot Blocking Buffer

Azure Fluorescent Blot Blocking Buffer

Blocking Buffer for Fluorescent Western blots, 500 mL
Catalog # 75794-864
Supplier:   Azure Biosystems
Supplier #:  AC2190
Azure Fluorescent Blot Blocking Buffer
Catalog # 75794-864
Supplier:  Azure Biosystems
CAS Number:  
Azure Fluorescent Blot Blocking Buffer
Azure Fluorescent Blot Blocking Buffer

Blocking Buffer for Fluorescent Western blots, 500 mL

CAS Number:  
Price
$309.93
Log in for availability
Each (500ml)
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Blocking buffer optimized for use with Azure’s fluorescent (and chemiluminescent) Western blotting systems.

Blocking Buffer for Fluorescent Western blots. 50x concentrated solution sufficient for 50-100 blots.

Specifications

Specifications

  • Description:
    Blocking Buffer for Fluorescent Western blots
  • Size:
    500 mL

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.