ContactUsLinkComponent
Manual stereotaxic injector, STOELTING MS
Manual stereotaxic injector, STOELTING MS
Catalog # 100231-302
Supplier:  Stoelting
Minority or Woman-Owned Business Enterprise
Manual stereotaxic injector, STOELTING MS
Catalog # 100231-302
Supplier:  Stoelting
Supplier Number:  51639
Minority or Woman-Owned Business Enterprise
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Volume:
    1 - 100 µl
  • Description:
    Manual Stereotaxic Injector
  • Cat. No.:
    100231-302
  • Supplier no.:
    51639
Specifications
About this item
Manual Stereotaxic Injector provides an economical means of injecting/withdrawing a calibrated volume with simple, precise manual control
Mounted directly on a stereotaxic frame, it allows precise injection directly through the needle into the tissue. Injection directly through a needle on the syringe avoids the compliance issues that arise when injections are forced through plastic tubing. Volumes as small as 200 picoliters may be infused or withdrawn.