TSK-GEL® Phenyl-5PW is most effiicient hydrophobic column to screen for the selectivity, retention and recovery of most biomolecules.
It features a 1000 Å pore size in a polymethacrylate base material, this column is stable in either acid or caustic cleaning regimens and provides excellent access to larger molecules with low diffusion coefficients.
Stainless steel is the standard column format for TSK-GEL® Phenyl-5PW. Glass and PEEK column housings are available for samples that may adsorb to stainless steel. The alternative column formats are also suitable for general applications.