Refinery Gas Standard #5, Restek
Catalog # 75795-218
Supplier: Restek
CAS Number:
Specifications
- Concentration:Mol
- Description:Refinery Gas Standard #5
- Size:4.6 L
- Shelf Life:One year
- Pressure:60 psig
- Cat. No.:75795-218
- Accuracy:Analytical accuracy: ±2%
- Tolerance:Blend tolerance: ±2%
- Supplier no.:34443
Specifications
About this item
Refinery gas standards are available in three mixes with varying C5 unsaturates or extended C6+ components.
- Cylinder: 4.6 L at 60 psig
- Mini-regulator designed specially for these standards is also available
Refinery gas standard #5 features 26 components.
Ordering information: Refinery Gas Standard #5 contains the following compounds: Hydrogen, 12.500 %; Argon, 1.000 %; Nitrogen, 37.200 %; Carbon monoxide, 1.000 %; Carbon dioxide, 3.000 %; Methane, 5.000 %; Ethane, 4.000 %; Ethylene, 2.000 %; Acetylene, 1.000 %; Propane, 6.000 %; Propylene, 3.000 %; Propadiene, 1.000 %; Isobutane, 5.000 %; N-Butane, 4.000 %; Isobutylene, 1.000 %; 1,3 Butadiene, 3.000 %; cis-2-Butene, 2.000 %; trans-2-Butene, 3.000 %; 1-Butene, 2.000 %; 2-Methyl-2-butene, 0.200 %; Isopentane, 1.000 %; N-Pentane, 1.000 %; cis-2-Pentene, 0.400 %; trans-2-Pentene, 0.200 %; Pentene-1, 0.400 %; Hexanes plus, 0.100 %.