Order Entry
Puerto Rico
ContactUsLinkComponent
Sulfur 5-Component Mix, Restek
Sulfur 5-Component Mix, Restek
Catalog # 75795-036
Supplier:  Restek
CAS Number:  
Sulfur 5-Component Mix, Restek
Catalog # 75795-036
Supplier:  Restek
Supplier Number:  34561
CAS Number:  

Specifications

  • Concentration:
    1 ppm in nitrogen
  • Description:
    Sulfur 5-Component Mix
  • Size:
    110 L
  • Shelf Life:
    One year
  • Pressure:
    1,800 psi
  • Cat. No.:
    75795-036
  • W×H:
    8.3×29.5 cm (3.25×11.62")
  • Accuracy:
    Analytical accuracy: ±10%
  • Tolerance:
    Blend tolerance: ±10%
  • Weight:
    1 kg (2.2 lbs.)
  • Supplier no.:
    34561

Specifications

About this item

Environmental Air Sampling Gas Standards meet lab requirements for two separate sources of calibration standards.

  • High-quality
  • No rental or demurrage fees required
  • Fast delivery provides freshest standard possible

Our high-quality air sampling gas calibration standards are provided by Spectra/Linde and Scott/Air Liquide—meeting lab requirements for two separate sources of calibration standards. Each comes with a certificate of analysis and unique serial number. All cylinders are disposable and do not require rental or demurrage fees. Recertification of cylinders is available directly with our suppliers. All cylinders are drop-shipped from our suppliers to provide fast delivery and the "freshest" standard possible. Minimum 12-month stability on all cylinders.

Ordering information: Sulfur-5 Component Mix consists of the following components: Carbonyl sulfide; Dimethyl sulfide; Ethyl mercaptan; Hydrogen sulfide; Methyl mercaptan. Stability is 12 months from date of manufacture. +/– 10% accuracy.