

4-Nitrobenzoic acid ≥99.0% (by HPLC), Sigma-Aldrich®
Catalog # EM8.00646.9050
Supplier: MilliporeSigma
CAS Number:
Specifications
- Pack type:Fibre carton
- Size:100 g
- Cat. No.:EM8.00646.9050
- Supplier no.:8.00646.9050
Specifications
About this item
:
:167.12 g/mol
Boiling Pt:359 °C (1013 hPa)
Melting Pt:239…242 °C
Density:1.61 g/cm3 (20 °C)
:M-Clarity™ quality level = MQ200
:GHS08
:GHS07
Storage Temperature:Ambient
CAS Number:62-23-7
Formula:C₇H₅NO₄
MDL Number:MFCD00007352