Order Entry
Puerto Rico
ContactUsLinkComponent
Tetraethylammonium hydroxide 20% in aqueous solution, Sigma-Aldrich®
Tetraethylammonium hydroxide 20% in aqueous solution, Sigma-Aldrich®
Catalog # EM8.22149.9025
Supplier:  MilliporeSigma
CAS Number:  
Tetraethylammonium hydroxide 20% in aqueous solution, Sigma-Aldrich®
Catalog # EM8.22149.9025
Supplier:  MilliporeSigma
Supplier Number:  8.22149.9025
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Barrel PE/metal
  • Size:
    25 kg
  • Cat. No.:
    EM8.22149.9025
  • Supplier no.:
    8.22149.9025

Specifications

About this item

 :
 :147.26
ADR:8,II
 :M-Clarity™ quality level = MQ200
UN:3267
CAS Number:77-98-5
Formula:C₈H₂₁NO
MDL Number:MFCD00009024