About this item
Recommended for small molecular weight compounds (<10000 Da) such as peptides, amino acids, tryptic digests, nucleotides, pharmaceutical molecules, and food and beverage samples.
- Provide superior resolution and speed
- Improved sensitivity
- Pressure drop is not excessive due to the monodisperse particle size distribution
TSKgel® Super-ODS, Super-Octyl and Super-Phenyl packings are bonded with, respectively, C18, C8 and phenyl functional groups. The bonded phases have a polymeric structure. An exhaustive endcapping reaction minimizes the presence of residual silanol groups.