ContactUsLinkComponent
Accessories for Vacuum Ovens, Shel Lab
  490012-640
Small Business Enterprise
Accessories for Vacuum Ovens, Shel Lab
  490012-640
 :  3450706
Small Business Enterprise
  • Description:
    Vacuum oven gasket, black silicone, 0.5 kg, 230 ºC
  • Cat. No.:
    490012-640
  • Supplier no.:
    3450706
This gasket is a replacement for both a 9.2 x 9.2 and 9.3 x 9.3 gasket.
  • General and high temperature applications
  • Resistant to moderate or oxidizing chemicals, ozone, concentrated sodium hydroxide
  • Attacked by many solvents, oils, concentrated acides, dilute sodium hydroxide