Orders LinkContactUsLinkComponent
TSK-Gel® NPH Columns, Tosoh Bioscience
Marketsource
Supplier:   Tosoh Bioscience
TSK-Gel® NPH Columns, Tosoh Bioscience
Supplier:  Tosoh Bioscience
Marketsource
Certificates
TSK-Gel® NPH Columns, Tosoh Bioscience
7 Options Available Below
Product Details & Documents
TSK-Gel® NPH columns are stainless steel with polymer based.
Specifications for Product Family
Specifications
  • Catalog No:
  • 78048-878
  • 78048-994
  • 100367-958
  • 100367-956
  • 78048-526
  • 78048-742
  • 78048-822
  • 78048-992
  • 78049-692
  • 100367-700
  • 100367-724
  • 100368-046
  • 100367-606
  • Max. Pressure:
  • 200 bar
  • pH Range:
  • 1.0 - 14.0
  • 2.0 - 12.0
  • 2.0 - 12.0
  • 2.0 - 12.0
  • 2.0 - 12.0
  • 2.0 - 12.0
  • 2.0 - 12.0
  • 1.0 - 14.0
  • 2.0 - 12.0
  • Temperature Limit:
  • 60 °C
Specifications
Product Family Options
Guard columns
Product Information
  • Column Format
    Phase
    Column hardware
    Particle Size
    Ø int.
    Length
    Packing material
    Availability
    Price
  • 78048-878
    TSK-Gel® DEAE-NPR Guard column
    TSK-Gel® DEAE-NPR Guard column
    Guard column
    DEAE
    Stainless Steel
    5 µm
    4.60 mm
    5 mm
    Silica
    Log in for availability
    Log in for availability
    Each
    Each - $623.58
    $623.58
    Each
       $623.58
    Supplier Number: 17088
    Specifications:
    New Product
    More
    Specifications
    Cat. No.
    78048-878
    Max. Pressure
    pH Range
    Temperature Limit
    New Product
  • 78048-994
    TSK-Gel® DNA-NPR Guard column
    TSK-Gel® DNA-NPR Guard column
    Guard column
    DEAE
    Stainless Steel
    5 µm
    4.60 mm
    5 mm
    Silica
    Log in for availability
    Log in for availability
    Each
    Each - $623.58
    $623.58
    Each
       $623.58
    Supplier Number: 18253
    Specifications:
    New Product
    More
    Specifications
    Cat. No.
    78048-994
    Max. Pressure
    pH Range
    Temperature Limit
    New Product
  • 100367-958
    TSK-Gel® DNA-NPR Guard column
    TSK-Gel® DNA-NPR Guard column
    Guard column
    DEAE
    Stainless Steel
    5 µm
    4.60 mm
    5 mm
    Silica
    Discontinued
    Supplier Number: 18253
    Specifications:
    Marketsource
    More
    Specifications
    Cat. No.
    100367-958
    Max. Pressure
    pH Range
    Temperature Limit
    Marketsource
  • 100367-956
    TSK-Gel® DEAE-NPR Guard column
    TSK-Gel® DEAE-NPR Guard column
    Guard column
    DEAE
    Stainless Steel
    5 µm
    4.60 mm
    5 mm
    Silica
    Discontinued
    Supplier Number: 17088
    Specifications:
    Marketsource
    More
    Specifications
    Cat. No.
    100367-956
    Max. Pressure
    pH Range
    Temperature Limit
    Marketsource
Product Information
  • Column Format
    Phase
    Column hardware
    Particle Size
    Ø int.
    Length
    Packing material
    Availability
    Price
  • 78048-526
    TSK-Gel® SAX
    TSK-Gel® SAX
    Analytical
    SAX (Strong Anion Exchange)
    Stainless Steel
    5 µm
    6.00 mm
    150 mm
    Silica
    Log in for availability
    Log in for availability
    Each
    Each - $1,798.37
    $1,798.37
    Each
       $1,798.37
    Supplier Number: 07157
    Specifications:
    New Product
    More
    Specifications
    Cat. No.
    78048-526
    Max. Pressure
    pH Range
    1.0 - 14.0
    Temperature Limit
    New Product
  • 78048-742
    TSK-Gel® DEAE-NPR, non porous
    TSK-Gel® DEAE-NPR, non porous
    Analytical
    DEAE
    Stainless Steel
    2.5 µm
    4.60 mm
    35 mm
    Silica
    Log in for availability
    Log in for availability
    Each
    Each - $1,819.03
    $1,819.03
    Each
       $1,819.03
    Supplier Number: 13075
    Specifications:
    New Product
    More
    Specifications
    Cat. No.
    78048-742
    Max. Pressure
    pH Range
    2.0 - 12.0
    Temperature Limit
    New Product
  • 78048-822
    TSK-Gel® Butyl-NPR, non porous
    TSK-Gel® Butyl-NPR, non porous
    Analytical
    Butyl
    Stainless Steel
    2.5 µm
    4.60 mm
    35 mm
    Silica
    Log in for availability
    Log in for availability
    Each
    Each - $1,870.22
    $1,870.22
    Each
       $1,870.22
    Supplier Number: 14947
    Specifications:
    New Product
    More
    Specifications
    Cat. No.
    78048-822
    Max. Pressure
    pH Range
    2.0 - 12.0
    Temperature Limit
    New Product
  • 78048-992
    TSK-Gel® DNA-NPR, non porous
    TSK-Gel® DNA-NPR, non porous
    Analytical
    DEAE
    Stainless Steel
    2.5 µm
    4.60 mm
    75 mm
    Silica
    Log in for availability
    Log in for availability
    Each
    Each - $2,473.62
    $2,473.62
    Each
       $2,473.62
    Supplier Number: 18249
    Specifications:
    New Product
    More
    Specifications
    Cat. No.
    78048-992
    Max. Pressure
    pH Range
    2.0 - 12.0
    Temperature Limit
    New Product
  • 78049-692
    TSKgel Butyl-NPR
    TSKgel Butyl-NPR
    Analytical
    Butyl
    Stainless Steel
    2.5 µm
    4.60 mm
    100 mm
    Silica
    Log in for availability
    Log in for availability
    Each
    Each - $2,387.16
    $2,387.16
    Each
       $2,387.16
    Supplier Number: 42168
    Specifications:
    New Product
    More
    Specifications
    Cat. No.
    78049-692
    Max. Pressure
    200 bar
    pH Range
    2.0 - 12.0
    Temperature Limit
    60 °C
    New Product
  • 100367-700
    TSK-Gel® DEAE-NPR, non porous
    TSK-Gel® DEAE-NPR, non porous
    Analytical
    DEAE
    Stainless Steel
    2.5 µm
    4.60 mm
    35 mm
    Silica
    Discontinued
    Supplier Number: 13075
    Specifications:
    Marketsource
    More
    Specifications
    Cat. No.
    100367-700
    Max. Pressure
    pH Range
    2.0 - 12.0
    Temperature Limit
    Marketsource
  • 100367-724
    TSK-Gel® Butyl-NPR, non porous
    TSK-Gel® Butyl-NPR, non porous
    Analytical
    C4 (Butyl)
    Stainless Steel
    2.5 µm
    4.60 mm
    35 mm
    Silica
    Discontinued
    Supplier Number: 14947
    Specifications:
    Marketsource
    More
    Specifications
    Cat. No.
    100367-724
    Max. Pressure
    pH Range
    2.0 - 12.0
    Temperature Limit
    Marketsource
  • 100368-046
    TSK-Gel® SAX
    TSK-Gel® SAX
    Analytical
    SAX (Strong Anion Exchange)
    Stainless Steel
    5 µm
    6.00 mm
    150 mm
    Silica
    Discontinued
    Supplier Number: 07157
    Specifications:
    Marketsource
    More
    Specifications
    Cat. No.
    100368-046
    Max. Pressure
    pH Range
    1.0 - 14.0
    Temperature Limit
    Marketsource
  • 100367-606
    TSK-Gel® DNA-NPR, non porous
    TSK-Gel® DNA-NPR, non porous
    Analytical
    DEAE
    Stainless Steel
    2.5 µm
    4.60 mm
    75 mm
    Silica
    Discontinued
    Supplier Number: 18249
    Specifications:
    Marketsource
    More
    Specifications
    Cat. No.
    100367-606
    Max. Pressure
    pH Range
    2.0 - 12.0
    Temperature Limit
    Marketsource