TSK-Gel® Reversed Phase Columns, Tosoh Bioscience
These silica- and polymer-based columns are ideal for reversed-phase applications such as pharmaceutical drugs, forensic compounds, derivatised amino acids, carbohydrates, steroids, lipids and fatty acids. Depending on their molecular weight, peptides may be analysed with the 80 to 140 Å silica-based columns, while proteins require a larger pore size column (200 to 300 Å).
- Silica and polymer-based materials
- Super series ideal for LCMS
- High recovery for biomolecules
TSKgel® Protein C4-300:
TSKgel® Protein C4-300 consists of a polymerically bonded butyl (C4) phase on 300 Å ultra-pure silica. The latest surface modification techniques and endcapping of residual silanol groups, reduce undesirable secondary interactions and peak tailing. It has been designed for the optimal recovery and resolution of biomolecules such as recombinant proteins, antibody fragments or PEGylated proteins.
TSKgel® ODS-140HTP:
TSKgel® ODS-140HTP columns were developed for use in high throughput (HTP) applications including drug discovery, pharmacokinetics and peptide digest separations. They are packed with 2.3 µm particles, providing high resolution and short analysis times at moderate pressure. The lower pressure drop enables these columns to be used with either UHPLC (up to 9006.84 psi) or conventional HPLC systems.
TSKgel® Super series:
The silica particles used in the Super series columns are monodisperse spherical 2.3 µm beads which are polymerically bonded and exhaustively end capped, to produce low metal-content, low silanol phases. The 2.3 µm particle provides superior resolution and speed of analysis as well as improved sensitivity. The columns do not suffer from excessive pressure drops.
Polymer-based phases:
The polymer-based reversed phase materials are chemically stable from pH 2 to 12, allowing operation at basic pH where silica-based phases have limited chemical stability. The non-porous Octadecyl-NPR column produces extremely fast kinetics and quantitative recovery of proteins at sub-microgram loads. TSKgel® Octadecyl-4PW is suitable for the analysis of peptides and small proteins, whereas the TSKgel® Octadecyl-2PW is used for small pharmaceutical compounds at basic pH. TSKgel® Phenyl-5PW has a high loading capacity and is ideal for the separation of high molecular weight proteins.
These silica- and polymer-based columns are ideal for reversed-phase applications such as pharmaceutical drugs, forensic compounds, derivatised amino acids, carbohydrates, steroids, lipids and fatty acids. Depending on their molecular weight, peptides may be analysed with the 80 to 140 Å silica-based columns, while proteins require a larger pore size column (200 to 300 Å).
- Silica and polymer-based materials
- Super series ideal for LCMS
- High recovery for biomolecules
TSKgel® Protein C4-300:
TSKgel® Protein C4-300 consists of a polymerically bonded butyl (C4) phase on 300 Å ultra-pure silica. The latest surface modification techniques and endcapping of residual silanol groups, reduce undesirable secondary interactions and peak tailing. It has been designed for the optimal recovery and resolution of biomolecules such as recombinant proteins, antibody fragments or PEGylated proteins.
TSKgel® ODS-140HTP:
TSKgel® ODS-140HTP columns were developed for use in high throughput (HTP) applications including drug discovery, pharmacokinetics and peptide digest separations. They are packed with 2.3 µm particles, providing high resolution and short analysis times at moderate pressure. The lower pressure drop enables these columns to be used with either UHPLC (up to 9006.84 psi) or conventional HPLC systems.
TSKgel® Super series:
The silica particles used in the Super series columns are monodisperse spherical 2.3 µm beads which are polymerically bonded and exhaustively end capped, to produce low metal-content, low silanol phases. The 2.3 µm particle provides superior resolution and speed of analysis as well as improved sensitivity. The columns do not suffer from excessive pressure drops.
Polymer-based phases:
The polymer-based reversed phase materials are chemically stable from pH 2 to 12, allowing operation at basic pH where silica-based phases have limited chemical stability. The non-porous Octadecyl-NPR column produces extremely fast kinetics and quantitative recovery of proteins at sub-microgram loads. TSKgel® Octadecyl-4PW is suitable for the analysis of peptides and small proteins, whereas the TSKgel® Octadecyl-2PW is used for small pharmaceutical compounds at basic pH. TSKgel® Phenyl-5PW has a high loading capacity and is ideal for the separation of high molecular weight proteins.
Product Family Options
Silica
- DescriptionPhasePore sizeColumn hardwareParticle SizeØ int.LengthPacking material
- TSKgel ODS-120TC18 (ODS or Octadecyl)120 ÅStainless Steel5 µm4.60 mm250 mmSilicaEach$912.97: 07125New ProductNew Product
- TSKgel TMS-250—250 ÅStainless Steel10 µm4.60 mm75 mmSilicaEach$1,376.34: 07190New ProductNew Product
- TSKgel ODS-120TC18 (ODS or Octadecyl)120 ÅStainless Steel5 µm4.60 mm150 mmSilicaEach$866.45: 07637New ProductNew Product
- TSKgel ODS-80TSC18 (ODS or Octadecyl)80 ÅStainless Steel5 µm2.00 mm150 mmSilicaEach$802.71: 18150New ProductNew Product
- TSKgel ODS-80TSC18 (ODS or Octadecyl)80 ÅStainless Steel5 µm2.00 mm250 mmSilicaEach$866.45: 18151New ProductNew Product
- TSKgel ODS-80TSC18 (ODS or Octadecyl)80 ÅStainless Steel5 µm4.60 mm150 mmSilicaEach$866.45: 17201New ProductNew Product
- TSKgel ODS-80TSC18 (ODS or Octadecyl)80 ÅStainless Steel5 µm4.60 mm250 mmSilicaEach$912.97: 17202New ProductNew Product
- TSKgel ODS-80TSC18 (ODS or Octadecyl)80 ÅStainless Steel5 µm2.00 mm150 mmSilica-: 18150MarketsourceMarketsource
- TSKgel ODS-80TSC18 (ODS or Octadecyl)80 ÅStainless Steel5 µm2.00 mm250 mmSilica-: 18151MarketsourceMarketsource
- TSKgel TMS-250—250 ÅStainless Steel10 µm4.60 mm75 mmSilica-: 07190MarketsourceMarketsource
- TSKgel ODS-120TC18 (ODS or Octadecyl)120 ÅStainless Steel5 µm4.60 mm150 mmSilica-: 07637MarketsourceMarketsource
- TSKgel ODS-80TSC18 (ODS or Octadecyl)80 ÅStainless Steel5 µm4.60 mm150 mmSilica-: 17201MarketsourceMarketsource
- TSKgel ODS-120TC18 (ODS or Octadecyl)120 ÅStainless Steel5 µm4.60 mm250 mmSilica-: 07125MarketsourceMarketsource
- TSKgel ODS-80TSC18 (ODS or Octadecyl)80 ÅStainless Steel5 µm4.60 mm250 mmSilica-: 17202MarketsourceMarketsource
Polymer
- DescriptionPhasePore sizeColumn hardwareParticle SizeØ int.LengthPacking material
- TSKgel Octadecyl-NPRC18 (ODS or Octadecyl)Stainless Steel2.5 µm4.60 mm35 mmSilicaEach$1,670.90: 14005New ProductNew Product
- TSKgel Phenyl-5PWPhenyl1000 ÅStainless Steel10 µm4.60 mm75 mmSilicaEach$1,131.74: 08043New ProductNew Product
- TSKgel Octadecyl-4PWC18 (ODS or Octadecyl)500 ÅStainless Steel7 µm4.60 mm150 mmSilicaEach$1,131.74: 13351New ProductNew Product
- TSKgel Octadecyl-2PWC18 (ODS or Octadecyl)100 - 200 ÅStainless Steel5 µm4.60 mm150 mmSilicaEach$1,267.80: 17500New ProductNew Product
- TSKgel Octadecyl-NPRC18 (ODS or Octadecyl)Stainless Steel2.5 µm4.60 mm35 mmSilica-: 14005MarketsourceMarketsource
- TSKgel Octadecyl-2PWC18 (ODS or Octadecyl)100 - 200 ÅStainless Steel5 µm4.60 mm150 mmSilica-: 17500MarketsourceMarketsource
- TSKgel Octadecyl-4PWC18 (ODS or Octadecyl)500 ÅStainless Steel7 µm4.60 mm150 mmSilica-: 13351MarketsourceMarketsource
- TSKgel Phenyl-5PWPhenyl1000 ÅStainless Steel10 µm4.60 mm75 mmSilica-: 08043MarketsourceMarketsource
Guard columns
- DescriptionPhasePore sizeColumn hardwareParticle SizeØ int.LengthPacking material
- Guard filter holderC18 (ODS or Octadecyl)Stainless Steel4.00 mm4 mmSilicaEach$318.67: 18206New ProductNew Product
- Guard filter, for 4.6 mm ID columns (Super-ODS, -Octyl, -Phenyl)C18 (ODS or Octadecyl)Stainless Steel2 µm4.00 mm4 mmSilicaEach$482.31: 18207New ProductNew Product
- Guard filter holderC18 (ODS or Octadecyl)Stainless Steel4.00 mm4 mmSilica-: 18206MarketsourceMarketsource
- Guard filter, for 4.6 mm ID columns (Super-ODS, -Octyl, -Phenyl)C18 (ODS or Octadecyl)Stainless Steel2 µm4.00 mm4 mmSilica-: 18207MarketsourceMarketsource
Guard Cartridge
- DescriptionPhasePore sizeColumn hardwareParticle SizeØ int.LengthPacking material
- TSKgel Octadecyl-4PW Guard cartridge, for 4.6 mm ID ColumnsStainless Steel7 µm3.20 mm15 mmEach$471.97: 19008New ProductNew Product
- TSKgel Octadecyl-4PW guard cartridge, for 4.6 mm ID columnsStainless Steel7 µm3.20 mm15 mm-: 19008MarketsourceMarketsource