KIMBLE® Graduated Test Tubes, with Flat Head Glass Stoppers, DWK Life Sciences
: Avantor
Manufactured from low extractable borosilicate glass.
- Graduated at 0.5 ml intervals
These round bottom test tubes have a solid flat head standard taper stopper.
: Conforming to USP Type I and ASTM E438, Type I, Class A requirements.