DNA Suspension Buffer, Teknova
Supplier: Teknova
Jump to:
About this item
RNAse, DNAse Tested (10mM Tris, 0.1mM EDTA)
4 Options Available Below
Product Details & Documents
RNAse, DNAse Tested (10mM Tris, 0.1mM EDTA)
Certificates
Search by Lot NumberProduct Family Options
Product Information
- SizeApplicationpHAvailabilityPrice
- Supplier Number: T0221
- 100 mLPCR, Ligation, Restriction Digestion, DNA Sequencing8.0Log in for availabilityEach (100ml)$79.76Supplier Number: T0227
- 250 mLPCR, Ligation, Restriction Digestion, DNA Sequencing8.0Log in for availabilityEach (250ml)$91.36Supplier Number: T0223
- 1 LPCR, Ligation, Restriction Digestion, DNA Sequencing8.0Log in for availabilityEach (1,000ml)$284.45Supplier Number: T0220