ContactUsLinkComponent
Corticosterone Human ELISA, Multispecies specificity, BioVendor
Corticosterone Human ELISA, Multispecies specificity, BioVendor
Catalog # 102552-156
Supplier:  BioVendor
CAS Number:  
undefined
Corticosterone Human ELISA, Multispecies specificity, BioVendor
Catalog # 102552-156
Supplier:  BioVendor
Supplier Number:  YK240
CAS Number:  
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Primary antibody reactivity:
    Human
  • Description:
    Corticosterone Human ELISA (Multispecies specificity)
  • Size:
    1 kit
  • Regulatory Status:
    RUO
  • Cat. No.:
    102552-156
  • Supplier no.:
    RSCYK240R
Specifications
About this item
The corticosterone EIA kit newly developed will be a quite useful tool for further development of corticosterone research.
  • Calibration Range: 0.21–50 ng/mL
  • Cell culture supernatant, Plasma, Serum, Urine
Corticosterone (C21H30O4, also called 11β,21-Dihydroxyprogesterone, Reichstein’s Substance H, or Kendall’s Compound B) is, like cortisol and cortisone, a glucocorticoid hormone secreted from the cortex of adrenal gland. Corticosterone is derived from cholesterol through a series of enzymatically mediated steps and also serving as a precursor of aldosterone. It is a primary glucocorticoid in mice and rats and other animals (such as rabbits, birds, amphibians, and reptilians) in which the 17α-hydroxylase is supposed not to exist in adrenal gland. Corticosterone is produced under the control of ACTH and the production has a circadian rhythm with peak levels in the latter portion of the day in nocturnal animals like rats and is believed to play a decisive role in sleep-wake cycles. Corticosterone can be used as a non-invasive biomarker of stress study through the collection of urine and feces to avoid corticosterone increase of blood levels which is caused by normal invasive methods. Corticosterone is also being studied in different fields such as impairment of long-term memory retrieval, chronic corticosterone elevation due to dietary restrictions and response to burn injuries etc. Since most of corticosterone in blood is bound to a plasma protein called corticosteronebinding globulin (CBG), the determination of blood corticosterone with presently available commercial assay kits requires an initial extraction procedure. On the other hand, the present assay kit for corticosterone newly developed by our laboratory provides a tool for direct determination of corticosterone in blood by simple dilution of blood samples with the diluent included in the kit. Furthermore, assays using the kit can be completed within a short period. The corticosterone EIA kit newly developed will be a quite useful tool for further development of corticosterone research.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.