ContactUsLinkComponent
Corticotropin Releasing Factor Mouse/Rat ELISA, BioVendor
  102552-158
 :  BioVendor
 :  
undefined
Corticotropin Releasing Factor Mouse/Rat ELISA, BioVendor
  102552-158
 :  BioVendor
 :  YK131
 :  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
  • Host:
  • Primary antibody reactivity:
    Mouse
  • Description:
    Corticotropin Releasing Factor Mouse/Rat ELISA
  • Size:
    1 kit
  • Environmentally Preferable:
  • Regulatory Status:
    RUO
  • Cat. No.:
    102552-158
  • Supplier no.:
    RSCYK131R
CRF serves as a potential neurochemical marker of early dementia and possible early AD.
  • Calibration Range: 0.078–2.5 ng/mL
  • Brain extract, Plasma-Citrate, Plasma-EDTA, Plasma-Heparin: 50 uL/well
Corticotropin releasing factor (CRF, also CRH) was initially isolated from ovine hypothalamus by Vale et al., in 1981, and identified as a novel neuropeptide comprising 41 amino acid residues with molecular weight 4758. Later human CRF and rat CRF were also isolated and identified. The mouse CRF peptide is identical at amino acid level to the rat and human CRF peptides. CRF in anterior pituitary promotes the synthesis and secretion of ACTH, a main factor of hypothalamus-pituitary-adrenal (HPA) axis. In the rat and human, CRF distributes mainly in hypothalamus, but it was also found in spinal cord, stomach, spleen, duodenum, adrenal and placenta. In addition, immunochemical evidence supported the wide distribution of the peptide throughout the central nervous system (CNS) such as olfactory bulb, retina and central auditory system in the rat. In mouse brain extracts, the highest concentrations of CRF-like immunoreactivity (CRF-LI) has been detected in the median eminence and hypothalamus and also existing in the amygdala, thalamus, frontal cortex, medulla/pons and cerebellum by radioimmunoassay. However because of the wide distribution, it is still disputing about CRF whether its blood level can reflect only the function of HPA axis. The relationships between CRF and stress, CRF and Alzheimer disease (AD) were attracted much attention recently. In fact the peptide was also suggested to regulate endocrine, autonomic and behavioral responses to stress, based on an experiment with acute and chronic stress rat models that showed endocrine function changes similar to those seen in patients with depression CRF in serial cerebrospinal fluid (CSF) of patients with depression was strikingly reduced as compared to those of normal subjects. The mean CRF and ACTH levels in the CSF of AD patients were significantly lower than those of healthy controls. Only in the cortices of those with mild dementia, CRF was reduced significantly, thus CRF was proposed to serve as a potential neurochemical marker of early dementia and possible early AD.
 : For research use only. Not for use in diagnostic or therapeutic procedures.