ContactUsLinkComponent
Glicentin Rat ELISA, BioVendor
Glicentin Rat ELISA, BioVendor
Catalog # 101424-320
Supplier:  BioVendor
CAS Number:  
undefined
Glicentin Rat ELISA, BioVendor
Catalog # 101424-320
Supplier:  BioVendor
Supplier Number:  YK111
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Glicentin Rat ELISA
  • Size:
    1 kit
  • Regulatory Status:
    RUO
  • Cat. No.:
    101424-320
  • Supplier no.:
    RSCYK111R
Specifications
About this item
Glicentin is a 69-amino-acid peptide containing glucagon and oxyntomodulin sequences in the molecule.
  • Calibration Range: 0.206–50 pmol/mL
  • Plasma-Citrate, Plasma-EDTA, Plasma-Heparin: 30 uL/well
Glicentin is a 69-amino-acid peptide containing glucagon and oxyntomodulin sequences in the molecule. It is suggested that glicentin and oxyntomodulin are produced in the intestinal L-cells and glucagon in A-cells in the pancreas, these peptides are derived from a common precursor by two different tissue-specific processing pathways. In 1983, the amino acid sequence of human glicentin was deduced by Bell et al. from the genomic sequence of human preproglucagon. Glicentin is a major form of gut glucagon-like immunoreactants (Gut GLIs). In mammalian small intestine, proglucagon is processed into glicentin, oxyntomodulin, and glucagon -like peptide 1(GLP-1) and glucagon -like peptide 2 (GLP-2). GLP-1(7–37) and GLP-1(7–36)amide have been isolated from the intestine and pancreas. It has been known that the GLP-1 sequence is well conserved between species in all mammals studied. Using synthetic peptides, several investigators have demonstrated that in contrast to GLP-1 (1–37), truncated GLP-1(7–36)amide and GLP-1(7–37) have several physiological effects. However, the physiological role of glicentin, a major gut glucagon, is still unclear. It has been known that the circulating level of plasma glicentin-like peptides increases significantly nutrient ingestion. Yanaihara institute Inc. has succeeded in developing a specific and convenient EIA kit for determination of rat glicentin in plasma.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.