ContactUsLinkComponent
Progranulin Mouse ELISA, BioVendor
Progranulin Mouse ELISA, BioVendor
Catalog # 102552-690
Supplier:  BioVendor
CAS Number:  
undefined
Progranulin Mouse ELISA, BioVendor
Catalog # 102552-690
Supplier:  BioVendor
Supplier Number:  RAG014R
CAS Number:  
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Primary antibody reactivity:
    Mouse
  • Description:
    Progranulin Mouse ELISA
  • Size:
    1 kit
  • Regulatory Status:
    RUO
  • Cat. No.:
    102552-690
  • Supplier no.:
    RAG014R
Specifications
About this item
Progranulin can restrain TNF mediated pro-inflammatory processes.
  • Calibration Range: 0.125–8 ng/mL
  • Limit of Detection: 60 pg/mL
  • Cell culture supernatant, Serum: sample diluiton will need to be determined, start at 1:1000 for serum
Progranulin is also known as Granulin Epithelin Precursor, Proepithelin or Acrogranin. It is a 68.5 kDa protein, consisting of 593 amino acids (inclusive Signalpeptid), which appears in vivo in strongly glycosylated form and therefore has a size of approximately 90 kDa. Progranulin has seven conserved domains, which are separated by linker sequences. By means of proteolytic cleavage, catalyzed by serine proteases like e.g. elastase, 6–25 kDa large fragments result, that are called Granulines or Epithelines. Progranulin is expressed and secreted in particular in strongly proliferating tissues such as adenoid tissue, spleen, skin epithelium, gastrointestinal mucous membranes, haematopoietic cells and in tumor cells. Until now no specific receptors, which would obtain the effect of Progranulin or the Granulines are known. Progranulin seems to be a factor, which affects the wound healing positively. In case of skin lesions the expression is increased in ceratinocytes, in macrophages and in neutrophile cells. Progranulin affects the wound healing indirectly by activation of macrophages and stimulation of angiogenesis in the damaged tissue. The physiological effects of Progranulin and Granulines are oppositional. Progranulin can restrain TNF mediated pro-inflammatory processes. On the other hand the Granulines seem to stimulate the secretion of pro-inflammatory cytokines. The influence of Progranulin on inflammatory processes could be shown also in arteriosclerotic plaques. Here Progranulin is expressed by smooth muscle cells and affects the migration of monocytes and smooth muscle cells. In the central nervous system Progranulin is expressed in microglia and neurons (in neocortical and hippocampal pyramid cells as well as in purkinje cells in the cerebellum). On mRNA level a clear increase of Progranulin expression could be shown during infections or injuries of the CNS, for example in mucopolysaccharidosis type I and IIIB, in viral inflammations of CNS, in amyotrophic lateral sclerosis and in Alzheimer's disease. Beyond that Progranulin seems to be of relevance in the development of sex specific differences during pre- and postnatal development and also for the neural plasticity in adults.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.