Order Entry
United States
ContactUsLinkComponent
IP-10 Mouse ELISA, BioVendor
IP-10 Mouse ELISA, BioVendor
Catalog # 102552-444
Supplier:  BioVendor
CAS Number:  
undefined
IP-10 Mouse ELISA, BioVendor
Catalog # 102552-444
Supplier:  BioVendor
Supplier Number:  RAF077R
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Host:
  • Primary antibody reactivity:
    Mouse
  • Target protein:
    IP-10 (CXCL10)
  • Description:
    IP-10 Mouse ELISA
  • Size:
    1 kit
  • Environmentally Preferable:
  • Regulatory Status:
    RUO
  • Cat. No.:
    102552-444
  • Supplier no.:
    RAF077R

Specifications

About this item

IP-10 (Interferon-gamma inducible Protein 10kDa) also known as CXCL10, is secreted by several cell types in response to IFNg and LPS.

  • Calibration Range: 7.8-500 pg/mL
  • Limit of Detection: 6.5 pg/mL
  • Cell culture supernatant, Plasma-Citrate, Plasma-EDTA, Serum: 50 uL/well

IP-10 (Interferon-gamma inducible Protein 10kDa) also known as CXCL10, is secreted by several cell types in response to IFNg and LPS. These cell types include monocytes, endothelial cells and fibroblasts.The gene for IP-10 is located on chromosome 4 in a cluster among several other cytokines and encodes a 98 amino acid precursor protein. IP-10 has been attributed to several roles, such as chemoattraction for monocytes and T cells (but not for neutrophils), inhibition of bone marrow colony formation and angiogenesis, promotion of T cells adhesion molecule expression. IP-10 shares a common receptor, CXCR3, with the chemokine MIG, but has been shown to play a distinct role in host defense in infections. IP-10 expression has been associated with HIV infection, is involved in inflammatory skin disease (6) and other allergic diseases; it appears in inflammation of the nervous system and in Alzheimer’s disease (astrocytes expressing IP-10 are commonly associated with senile plaques).

Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.