Order Entry
Puerto Rico
ContactUsLinkComponent
Interleukin-17, IL-17, Mouse ELISA, BioVendor
Interleukin-17, IL-17, Mouse ELISA, BioVendor
Catalog # 102552-324
Supplier:  BioVendor
CAS Number:  
undefined
Interleukin-17, IL-17, Mouse ELISA, BioVendor
Catalog # 102552-324
Supplier:  BioVendor
Supplier Number:  RAF038R
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Primary antibody reactivity:
    Mouse
  • Target protein:
    IL-17
  • Description:
    Interleukin-17 (IL-17) Mouse ELISA
  • Size:
    1 kit
  • Regulatory Status:
    RUO
  • Cat. No.:
    102552-324
  • Supplier no.:
    RAF038R

Specifications

About this item

Interleukin-17 is involved in the regulation of hematopoiesis and its effects are dependent on the physiological/pathological status of the organism.

  • Calibration Range: 7.8-500 pg/mL
  • Limit of Detection: 1.6 pg/mL
  • Cell culture supernatant, Serum: 50 uL/well

Interleukin-17 is involved in the regulation of hematopoiesis and its effects are dependent on the physiological/pathological status of the organism. IL-17 expression in airways is upregulated upon allergen exposure such as in allergic asthma. Its crucial role in T-cell activation suggests that IL-17 is involved in the development of autoimmune arthritis.

Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.