Order Entry
Puerto Rico
ContactUsLinkComponent
Interleukin-12/Interleukin-23 p40 Human ELISA, BioVendor
Interleukin-12/Interleukin-23 p40 Human ELISA, BioVendor
Catalog # 102552-316
Supplier:  BioVendor
CAS Number:  
Interleukin-12/Interleukin-23 p40 Human ELISA, BioVendor
Catalog # 102552-316
Supplier:  BioVendor
Supplier Number:  RAF034R
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Assay duration:
    Multiple steps
  • Assay Type (ELISA with LOV):
    Sandwich
  • Conjugate ELISA:
    Biotin
  • Format:
    Pre-coated
  • Primary antibody reactivity:
    Human
  • Target protein:
    IL-12 subunit p40, cytotoxic lymphocyte maturation factor 40 kDa subunit, NK cell stimulatory factor chain 2, IL-12B, IL-23, IL-23 subunit p40, T-cell stimulating factor
  • Description:
    Interleukin-12/Interleukin-23 p40 human ELISA
  • Size:
    1 kit
  • Sample Type:
    Serum, plasma, cell culture supernatant
  • Detection Method:
    Colorimetric
  • Shelf Life:
    See expiration date
  • Detection Range:
    31.3 - 2000 pg/ml
  • Storage Temperature:
    2…8 °C
  • Sensitivity:
    <10 pg/ml
  • Regulatory Status:
    RUO
  • Cat. No.:
    102552-316
  • Supplier no.:
    RAF034R

Specifications

About this item

96 wells (1 kit) - ELISA is a competitive enzyme immunoassay for the quantitative measurement of Interleukin-12/Interleukin-23 p40.

  • Calibration Range: 31.3-2000 pg/ml
  • Limit of Detection: < 10 pg/ml
  • Cell culture supernatant, Plasma, Serum: 50 ul/well

The human IL-12/IL-23 p40 ELISA is an enzyme-linked immunosorbent assay for the quantitative detection of human IL-12/IL-23 p40. The human IL-12/IL-23 p40 ELISA is for research use only. Not for diagnostic or therapeutic procedures.

IL-23 and IL-12 are heterodimeric cytokines which share the p40 subunit, but have unique second subunits, IL-23p19 and IL-12p35.

An anti-human IL-12/IL-23 p40 coating antibody is adsorbed onto microwells. Human IL-12/IL-23 p40 present in the sample or standard binds to antibodies adsorbed to the microwells. A biotin-conjugated anti-human IL-12/IL23 p40 antibody is added and binds to human IL12/IL-23 p40 captured by the first antibody. Following incubation unbound biotin-conjugated antihuman IL-12/IL-23 p40 antibody is removed during a wash step. Streptavidin-HRP is added and binds to the biotin-conjugated anti-human IL-12/IL-23 p40 antibody. Following incubation unbound Streptavidin-HRP is removed during a wash step, and substrate solution reactive with HRP is added to the wells. A colored product is formed in proportion to the amount of human IL-12/IL-23 p40 present in the sample or standard. The reaction is terminated by addition of acid and absorbance is measured at 450 nm. A standard curve is prepared from 7 human IL-12/IL-23 p40 standard dilutions and human IL12/IL-23 p40 sample concentration determined.

Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.
Caution: This is intended for research use only.