Hand Crimper, PerkinElmer
Catalog # 97013-494
Supplier: PerkinElmer
Specifications
- Description:Hand Crimper
- Environmentally Preferable:
- Cat. No.:97013-494
- Seal size:20 mm
- Supplier no.:N9302785
Specifications
About this item
Hand crimper for GC Headspace vials or LC large volume vials.
- Suited for large-volume vials
- Guarantees sample integrity
- Useful when handling volatile compounds
It ensures sample integrity, particularly those containing volatile compounds.