Heating Mantle Controllers, Chemglass
Catalog # 80061-222
Supplier: Chemglass
Small Business Enterprise
Specifications
- Description:OptiChem® Heating Mantle Controller, Single Circuit, 10A, 1200 Watts, 120VAC
- Cat. No.:80061-222
- Supplier no.:CG-15005-01
Specifications
About this item
These controllers feature solid-state proportional voltage power control in a compact benchtop design
They provide accurate manual control of heating mantles, tapes and all other resistive loads with an adjustable output range from 5 to 100% of the rated voltage. 3 year warranty.
Delivery: All units are supplied with a 6ft separable power cord, a grounded, 3-wire receptacle and lighted power switch.