Order Entry
Puerto Rico
ContactUsLinkComponent
Thermostable Dutpase (from Escherichia coli)
Thermostable Dutpase (from Escherichia coli)
Catalog # 101423-948
Supplier:  BioVendor
CAS Number:  
undefined
Thermostable Dutpase (from Escherichia coli)
Catalog # 101423-948
Supplier:  BioVendor
Supplier Number:  RP179281
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Source:
    E. coli
  • Size:
    1000 IU
  • Enzyme Name:
    Thermostable Dutpase
  • Enzyme Synonyms:
    dUTPase
  • Cat. No.:
    101423-948
  • Supplier no.:
    RP179281

Specifications

About this item

Thermostable dUTPase (pyrococcus fruriosus) maximizes the efficiency of high-fidelity PCR (using proofreading DNA polymerases)

It removes contaminating dUTP present in PCR reactions and dNTP solutions. The presence of dUTPase in a proofreading DNA polymerase reaction can prevent dUTP misincorporation by maintaining dUTP levels below their inhibitory concentrations despite the constant generation of the molecule by the spontaneous deamination of dCTP. The incorporation of dUTP into PCR products causes mutations within the amplified product, proofreading polymerases to stall and slows down non-proofreading polymerases such as Taq. The dUTPase increase in PCR product yield, length and fidelity enables further down-stream applications. These effects make dUTPase useful in PCR fidelity and yield-sensitive applications such as cloning and subsequent recombinant protein technology, and gene expression analysis (semi-quantitative RT-PCR techniques and real-time PCR analysis), where small differences in product accumulation can have a significant impact on gene expression analysis. dUTPase is specific for dUTP and is critical for the fidelity of DNA replication and repair. dUTPase hydrolyzes dUTP to dUMP and pyrophosphate, simultaneously reducing dUTP levels and providing the dUMP for dTTP biosynthesis.

Ordering information: For research use only.