Order Entry
United States
ContactUsLinkComponent
 
Human Recombinant Secretoglobin Family 3A (from E. coli)
  102830-692
 :  BioVendor
 :  
undefined
Human Recombinant Secretoglobin Family 3A (from E. coli)
  102830-692
 :  BioVendor
 :  RD172373100
 :  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

 

  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Size:
    100 µg
  • Environmentally Preferable:
  • Protein Synonyms:
    Uteroglobin-related protein 1|SCGB3A2|Pneumo secretory protein 1|PnSP-1
  • Protein/Peptide Name:
    Secretoglobin Family 3A
  • Sequence:
    MFLINKVPLP VDKLAPLPLD NILPFMDPLK LLLKTLGISV EHLVEGLRKC VNELGPEASE AVKKLLEALS HLVGGSGGSF LINKVPLPVD KLAPLPLDNI LPFMDPLKLL LKTLGISVEH LVEGLRKCVN ELGPEASEAV KKLLEALSHL VHHHHHH
  • Cat. No.:
    102830-692
  • Supplier no.:
    RD172373100

 

 

Secretoglobins are small disulfide linked dimeric proteins. Secretoglobin family 3A member 2 is highly expressed in lung.

Sequence: MFLINKVPLP VDKLAPLPLD NILPFMDPLK LLLKTLGISV EHLVEGLRKC VNELGPEASE AVKKLLEALS HLVGGSGGSF LINKVPLPVD KLAPLPLDNI LPFMDPLKLL LKTLGISVEH LVEGLRKCVN ELGPEASEAV KKLLEALSHL VHHHHHH

Other names: Pneumo secretory protein 1, PnSP-1, Uteroglobin-related protein 1, SCGB3A2

 : For research use only.