Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Human
- Size:100 µg
- Protein Synonyms:NPHS2
- Protein/Peptide Name:Podocin
- Molecular Weight:30.2 kDa
- Sequence:MKHHHHHHAS VKVVQEYERV IIFRLGHLLP GRAKGPGLFF FLPCLDTYHK VDLRLQTLEI PFHEIVTKDM FIMEIDAICY YRMENASLLL SSLAHVSKAV QFLVQTTMKR LLAHRSLTEI LLERKSIAQD AKVALDSVTC IWGIKVERIE IKDVRLPAGL QHSLAVEAEA QRQAKVRMIA AEAEKAASES LRMAAEILSG TPAAVQLRYL HTLQSLSTEK PSTVVLPLPF DLLNCLSSPS NRTQGSLPFP SPSKPVEPLN PKKKDSPML
- Cat. No.:102830-644
- Supplier no.:RD172330100
Specifications
About this item
Total 269 AA. MW: 30.2 kDa (calculated). UniProtKB acc.no. Q9NP85 (Val125-Leu383). N-terminal His-tag (10 extra AA). Protein identity confirmed by LC-MS/MS.
Sequence: MKHHHHHHAS VKVVQEYERV IIFRLGHLLP GRAKGPGLFF FLPCLDTYHK VDLRLQTLEI PFHEIVTKDM FIMEIDAICY YRMENASLLL SSLAHVSKAV QFLVQTTMKR LLAHRSLTEI LLERKSIAQD AKVALDSVTC IWGIKVERIE IKDVRLPAGL QHSLAVEAEA QRQAKVRMIA AEAEKAASES LRMAAEILSG TPAAVQLRYL HTLQSLSTEK PSTVVLPLPF DLLNCLSSPS NRTQGSLPFP SPSKPVEPLN PKKKDSPML
Other names: NPHS2