Order Entry
Puerto Rico
ContactUsLinkComponent
Human Recombinant alpha-Dg-N (from E. coli)
Human Recombinant alpha-Dg-N (from E. coli)
Catalog # 101189-688
Supplier:  BioVendor
CAS Number:  
undefined
Human Recombinant alpha-Dg-N (from E. coli)
Catalog # 101189-688
Supplier:  BioVendor
Supplier Number:  RD172318100
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Size:
    100 µg
  • Protein Synonyms:
    alpha-DG-N|Dystrophin-associated glycoprotein 1|DAG1
  • Protein/Peptide Name:
    alpha-Dg-N
  • Purity:
    >90%
  • Sequence:
    MKHHHHHHAS HWPSEPSEAV RDWENQLEAS MHSVLSDLHE AVPTVVGIPD GTAVVGRSFR VTIPTDLIAS SGDIIKVSGKEALPSWLH WDSQSHTLEG LPLDTDKGVH YISVSATRLG ANGSHIPQTS SVFSIEVYPE DHSELQSVRT ASPDPGEVVS SACAADEPVT VLTVILDADL TKMTPKQRID LLHRMRSFSE VELHNMKLVP VVNNRLFDMS AFMAGPGNAK KVVENGALLS WKLGCSLNQN SVPDIHGVEA PAREGAMSAQ LGYPVVGWHI ANKKPPLPKR VRR
  • Cat. No.:
    101189-688
  • Supplier no.:
    RD172318100

Specifications

About this item

N-terminal domain of alpha-dystroglycan is secreted into the cerebrospinal fluid. Its effect on the nervous system remains unclear. The complete dystroglycan complex is expressed in a variety of tissues and plays a role in processes such as laminin and basement membrane assembly, sarcolemmal stability, cell survival, peripheral nerve myelination, nodal structure, cell migration, and epithelial polarization.

Sequence: MKHHHHHHAS HWPSEPSEAV RDWENQLEAS MHSVLSDLHE AVPTVVGIPD GTAVVGRSFR VTIPTDLIAS SGDIIKVSAA GKEALPSWLH WDSQSHTLEG LPLDTDKGVH YISVSATRLG ANGSHIPQTS SVFSIEVYPE DHSELQSVRT ASPDPGEVVS SACAADEPVT VLTVILDADL TKMTPKQRID LLHRMRSFSE VELHNMKLVP VVNNRLFDMS AFMAGPGNAK KVVENGALLS WKLGCSLNQN SVPDIHGVEA PAREGAMSAQ LGYPVVGWHI ANKKPPLPKR VRR

Other names: Dystrophin-associated glycoprotein 1, DAG1, alpha-DG-N

Ordering information: For research use only.