Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Human
- Size:100 µg
- Protein Synonyms:FAM3CILEI|Interleukin-like EMT inducer|GS3786
- Protein/Peptide Name:FAM3C
- Purity:> 95%
- Sequence:MKHHHHHHAS QVFEIKMDAS LGNLFARSAL DTAARSTKPP RYKCGISKAC PEKHFAFKMA SGAANVVGPK ICLEDNVLMS GVKNNVGRGI NVALANGKTG EVLDTKYFDM WGGDVAPFIE FLKAIQDGTI VLMGTYDDGA TKLNDEARRL IADLGSTSIT NLGFRDNWVF CGGKGIKTKS PFEQHIKNNK DTNKYEGWPE VVEMEGCIPQ KQD
- Cat. No.:101189-674
- Supplier no.:RD172302100
FAM3C is a cytokine protein. It is involved in retinal laminar formation. It promotes epithelial to mesenchymal transition. FAM3C is overexpressed and mislocalized in multiple human tumors.
Sequence: MKHHHHHHAS QVFEIKMDAS LGNLFARSAL DTAARSTKPP RYKCGISKAC PEKHFAFKMA SGAANVVGPK ICLEDNVLMS GVKNNVGRGI NVALANGKTG EVLDTKYFDM WGGDVAPFIE FLKAIQDGTI VLMGTYDDGA TKLNDEARRL IADLGSTSIT NLGFRDNWVF CGGKGIKTKS PFEQHIKNNK DTNKYEGWPE VVEMEGCIPQ KQD
Other names: Interleukin-like EMT inducer, FAM3CILEI, GS3786