Order Entry
Puerto Rico
ContactUsLinkComponent
Human Recombinant Gastrokine-2 (from E. coli)
Human Recombinant Gastrokine-2 (from E. coli)
Catalog # 101174-836
Supplier:  BioVendor
CAS Number:  
undefined
Human Recombinant Gastrokine-2 (from E. coli)
Catalog # 101174-836
Supplier:  BioVendor
Supplier Number:  RD172269100
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Size:
    100 µg
  • Protein Synonyms:
    BLOT|GDDR|UNQ465/PRO813|TFIZ1|Blottin|GKN2
  • Protein/Peptide Name:
    Gastrokine-2
  • Purity:
    >90%
  • Sequence:
    MKHHHHHHAS YEVFNIISPS NNGGNVQETV TIDNEKNTAI INIHAGSCSS TTIFDYKHGY IASRVLSRRA CFILKMDHQN IPPLNNLQWY IYEKQALDNM FSSKYTWVKY NPLESLIKDV DWFLLGSPIE KLCKHIPLYK GEVVENTHNV GAGGCAKAGL LGILGISICA DIHV
  • Cat. No.:
    101174-836
  • Supplier no.:
    RD172269100

Specifications

About this item

Gastrokine-2 is expressed in gastric mucosa.

Sequence: MKHHHHHHAS YEVFNIISPS NNGGNVQETV TIDNEKNTAI INIHAGSCSS TTIFDYKHGY IASRVLSRRA CFILKMDHQN IPPLNNLQWY IYEKQALDNM FSSKYTWVKY NPLESLIKDV DWFLLGSPIE KLCKHIPLYK GEVVENTHNV GAGGCAKAGL LGILGISICA DIHV

Other names: Blottin, GKN2, BLOT, GDDR, TFIZ1, UNQ465/PRO813

Ordering information: For research use only.