Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Human
- Size:100 µg
- Protein Synonyms:AMP-18|GKN1|AMP18|CA11|18 kDa antrum mucosa protein|UNQ489/PRO1005
- Protein/Peptide Name:Gastrokine-1
- Purity:>90%
- Molecular Weight:18 kDa
- Sequence:MKHHHHHHAS NYNINVNDDN NNAGSGQQSV SVNNEHNVAN VDNNNGWDSW NSIWDYGNGF AATRLFQKKT CIVHKMNKEV MPSIQSLDAL VKEKKLQGKG PGGPPPKGLM YSVNPNKVDD LSKFGKNIAN MCRGIPTYMA EEMQEASLFF YSGTCYTTSV LWIVDISFCG DTVEN
- Cat. No.:101169-496
- Supplier no.:RD172268100
Gastrokine 1, previously known as AMP-18, CA11, FOVEOLIN and TFIZ, was formally named “GKN1” by the HUGO gene Nomenclature Committee for its gastric-specific expression and its highly conserved presence in the gastric mucosa of many mammalian species. Gastrokine 1 is a novel protein that was firstly cloned by a Japanese group in 2000. GKN1 belongs to a family of genes encoding stomach-specific secreted proteins consisting of 3 known members: gastrokine 1 (GKN1), gastrokine 2 (GKN2) and gastrokine 3 (GKN3). GKN1 gene of about 6 kb was reported to be located at 2p13 and contains 6 exons. The GKN1 gene encodes a small protein of 185 amino acids containing a N-terminal signal peptide. It is a secreted protein with a molecular weight of approximately 18 kDa. GKN1 protein contains a BRICHOS domain, which is associated with dementia, respiratory distress and cancer.
Sequence: MKHHHHHHAS NYNINVNDDN NNAGSGQQSV SVNNEHNVAN VDNNNGWDSW NSIWDYGNGF AATRLFQKKT CIVHKMNKEV MPSIQSLDAL VKEKKLQGKG PGGPPPKGLM YSVNPNKVDD LSKFGKNIAN MCRGIPTYMA EEMQEASLFF YSGTCYTTSV LWIVDISFCG DTVEN
Other names: 18 kDa antrum mucosa protein, AMP-18, CA11, GKN1, AMP18, UNQ489/PRO1005