Order Entry
Puerto Rico
ContactUsLinkComponent
 
Human Recombinant Renin Receptor (from E. coli)
Catalog # 101169-490
Supplier:  BioVendor
CAS Number:  
undefined
Human Recombinant Renin Receptor (from E. coli)
Catalog # 101169-490
Supplier:  BioVendor
Supplier Number:  RD172264100
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Size:
    100 µg
  • Protein Synonyms:
    Renin/prorenin|N14F|ER-localized type I transmembrane adaptor|ATPase H(+-transporting lysosomal-interacting protein 2|Embryonic liver differentiation factor 10|ATPase H(+-transporting lysosomal accessory protein 2
  • Protein/Peptide Name:
    Renin Receptor
  • Molecular Weight:
    33 kDa
  • Sequence:
    MKHHHHHHAS NEFSILKSPG SVVFRNGNWP IPGERIPDVA ALSMGFSVKE DLSWPGLAVG NLFHRPRATV MVMVKGVNKL ALPPGSVISY PLENAVPFSL DSVANSIHSL FSEETPVVLQ LAPSEERVYM VGKANSVFED LSVTLRQLRN RLFQENSVLS SLPLNSLSRN NEVDLLFLSE LQVLHDISSL LSRHKHLAKD HSPDLYSLEL AGLDEIGKRY GEDSEQFRDA SKILVDALQK FADDMYSLYG GNAVVELVTV KSFDTSLIRK TRTILEAKQA KNPASPYNLA YKYNFE
  • Cat. No.:
    101169-490
  • Supplier no.:
    RD172264100

Specifications

About this item

Total 296 AA. MW: 33 kDa (calculated). N-Terminal His-tag (10 extra AA). UniprotKB acc.no. O75787

Sequence: MKHHHHHHAS NEFSILKSPG SVVFRNGNWP IPGERIPDVA ALSMGFSVKE DLSWPGLAVG NLFHRPRATV MVMVKGVNKL ALPPGSVISY PLENAVPFSL DSVANSIHSL FSEETPVVLQ LAPSEERVYM VGKANSVFED LSVTLRQLRN RLFQENSVLS SLPLNSLSRN NEVDLLFLSE LQVLHDISSL LSRHKHLAKD HSPDLYSLEL AGLDEIGKRY GEDSEQFRDA SKILVDALQK FADDMYSLYG GNAVVELVTV KSFDTSLIRK TRTILEAKQA KNPASPYNLA YKYNFE

Other names: ATPase H(+)-transporting lysosomal accessory protein 2, ATPase H(+)-transporting lysosomal-interacting protein 2, ER-localized type I transmembrane adaptor, Embryonic liver differentiation factor 10, N14F, Renin/prorenin

Ordering information: For research use only.