Order Entry
United States
ContactUsLinkComponent
Human Gal-13 (from E. coli)
Human Gal-13 (from E. coli)
Catalog # 102552-934
Supplier:  BioVendor
CAS Number:  
undefined
Human Gal-13 (from E. coli)
Catalog # 102552-934
Supplier:  BioVendor
Supplier Number:  RD172254100
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Source:
    E. coli
  • Species:
    Human
  • Size:
    100 µg
  • Environmentally Preferable:
  • Protein Synonyms:
    PLAC8|Galactoside-Binding Soluble Lectin 13|Gal-13|PP13|Galectin-13|Placental tissue protein 13|LGALS13
  • Protein/Peptide Name:
    Gal-13
  • Sequence:
    MKHHHHHHAS MSSLPVPYKL PVSLSVGSCV IIKGTPIHSF INDPQLQVDF YTDMDEDSDI AFRFRVHFGN HVVMNRREFG IWMLEETTDY VPFEDGKQFE LCIYVHYNEY EIKVNGIRIY GFVHRIPPSF VKMVQVSRDI SLTSVCVCN
  • Cat. No.:
    102552-934
  • Supplier no.:
    RD172254100

Specifications

About this item

Placental protein 13 (PP13, Galectin 13) is the member of the beta-galactoside binding S-type galectin superfamily, whose members are important in placenta implantation and remodelling of maternal arteries. PP13 binds to beta-galactoside residues of several proteins on the cell surface, cytoskeleton and extracellular matrix, thereby generating various responses such as immune responses and influencing other functions like apoptosis and molecular recognition. PP13, which is predominantly produced by placental tissue, possesses a conserved carbohydrate binding domain, to which two proteins Annexin-II and Actin-beta bind. These proteins are considered to play a key role in placentation and maternal artery remodelling respectively.

Sequence: MKHHHHHHAS MSSLPVPYKL PVSLSVGSCV IIKGTPIHSF INDPQLQVDF YTDMDEDSDI AFRFRVHFGN HVVMNRREFG IWMLEETTDY VPFEDGKQFE LCIYVHYNEY EIKVNGIRIY GFVHRIPPSF VKMVQVSRDI SLTSVCVCN

Other names: Galactoside-Binding Soluble Lectin 13, Galectin-13, Gal-13, Placental tissue protein 13, PP13, LGALS13, PLAC8

Ordering information: For research use only.