Order Entry
Puerto Rico
ContactUsLinkComponent
Human Testis FABP (from E. coli)
Human Testis FABP (from E. coli)
Catalog # 102552-930
Supplier:  BioVendor
CAS Number:  
undefined
Human Testis FABP (from E. coli)
Catalog # 102552-930
Supplier:  BioVendor
Supplier Number:  RD172251100
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Source:
    E. coli
  • Species:
    Human
  • Size:
    100 µg
  • Protein Synonyms:
    Testis lipid-binding protein|FABP9|T-FABP|TLBP|Fatty acid-binding protein 9
  • Protein/Peptide Name:
    Testis FABP
  • Molecular Weight:
    16.2 kDa
  • Sequence:
    MKHHHHHHAS VEPFLGTWKL VSSENFEDYM KELGVNFAAR NMAGLVKPTV TISVDGKMMT IRTESSFQDT KISFKLGEEF DETTADNRKV KSTITLENGS MIHVQKWLGK ETTIKRKIVD EKMVVECKMN NIVSTRIYEK V
  • Cat. No.:
    102552-930
  • Supplier no.:
    RD172251100

Specifications

About this item

Total 141 AA. MW: 16.2 kDa (calculated). UniProtKB acc.no. Q0Z7S8. N-Terminal His-tag (10 extra AA)

Sequence: MKHHHHHHAS VEPFLGTWKL VSSENFEDYM KELGVNFAAR NMAGLVKPTV TISVDGKMMT IRTESSFQDT KISFKLGEEF DETTADNRKV KSTITLENGS MIHVQKWLGK ETTIKRKIVD EKMVVECKMN NIVSTRIYEK V

Other names: Testis lipid-binding protein, TLBP, Fatty acid-binding protein 9, T-FABP, FABP9

Ordering information: For research use only.