Order Entry
Puerto Rico
ContactUsLinkComponent
Human Recombinant Prohibitin (from E. coli)
Human Recombinant Prohibitin (from E. coli)
Catalog # 101169-486
Supplier:  BioVendor
CAS Number:  
undefined
Human Recombinant Prohibitin (from E. coli)
Catalog # 101169-486
Supplier:  BioVendor
Supplier Number:  RD172239100
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Size:
    100 µg
  • Protein Synonyms:
    PHB
  • Protein/Peptide Name:
    Prohibitin
  • Purity:
    >90 %
  • Sequence:
    MKHHHHHHAS MAAKVFESIG KFGLALAVAG GVVNSALYNV DAGHRAVIFD RFRGVQDIVV GEGTHFLIPW VQKPIIFDCR SRPRNVPVIT GSKDLQNVNI TLRILFRPVA SQLPRIFTSI GEDYDERVLP SITTEILKSV VARFDAGELI TQRELVSRQV SDDLTERAAT FGLILDDVSL THLTFGKEFT EAVEAKQVAQ QEAERARFVV EKAEQQKKIISAEGDSKA AELIANSLAT AGDGLIELRK LEAAEDIAYQ LSRSRNITYL PAGQSVLLQL PQ
  • Cat. No.:
    101169-486
  • Supplier no.:
    RD172239100

Specifications

About this item

Prohibitin is a mitochondrial protein. It restrains DNA synthesis and takes part in proliferation regulation. It may play a role in aging and regulating mitochondrial respiration activity.

Sequence: MKHHHHHHAS MAAKVFESIG KFGLALAVAG GVVNSALYNV DAGHRAVIFD RFRGVQDIVV GEGTHFLIPW VQKPIIFDCR SRPRNVPVIT GSKDLQNVNI TLRILFRPVA SQLPRIFTSI GEDYDERVLP SITTEILKSV VARFDAGELI TQRELVSRQV SDDLTERAAT FGLILDDVSL THLTFGKEFT EAVEAKQVAQ QEAERARFVV EKAEQQKKAA IISAEGDSKA AELIANSLAT AGDGLIELRK LEAAEDIAYQ LSRSRNITYL PAGQSVLLQL PQ

Other names: PHB

Ordering information: For research use only.