Order Entry
Puerto Rico
ContactUsLinkComponent
Human S100A5 (fromE. coli)
Human S100A5 (fromE. coli)
Catalog # 102552-876
Supplier:  BioVendor
CAS Number:  
undefined
Human S100A5 (fromE. coli)
Catalog # 102552-876
Supplier:  BioVendor
Supplier Number:  RD172212100
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Source:
    E. coli
  • Species:
    Human
  • Size:
    100 µg
  • Protein Synonyms:
    Protein S-100D|S100 calcium-binding protein A5
  • Protein/Peptide Name:
    S100A5
  • Molecular Weight:
    11.99 kDa
  • Sequence:
    MKHHHHHHAS METPLEKALT TMVTTFHKYS GREGSKLTLS RKELKELIKK ELCLGEMKES SIDDLMKSLD KNSDQEIDFK EYSVFLTMLC MAYNDFFLED NK
  • Cat. No.:
    102552-876
  • Supplier no.:
    RD172212100

Specifications

About this item

Total 102 AA. MW: 11.99 kDa (calculated). UniProtKB acc.no. P33763. N-Terminal His-tag, 10 extra AA

Sequence: MKHHHHHHAS METPLEKALT TMVTTFHKYS GREGSKLTLS RKELKELIKK ELCLGEMKES SIDDLMKSLD KNSDQEIDFK EYSVFLTMLC MAYNDFFLED NK

Other names: S100 calcium-binding protein A5, Protein S-100D

Ordering information: For research use only.