Order Entry
Puerto Rico
ContactUsLinkComponent
Human IL33 (from E. coli)
Human IL33 (from E. coli)
Catalog # 102552-864
Supplier:  BioVendor
CAS Number:  
undefined
Human IL33 (from E. coli)
Catalog # 102552-864
Supplier:  BioVendor
Supplier Number:  RD172203100
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Source:
    E. coli
  • Species:
    Human
  • Size:
    100 µg
  • Protein Synonyms:
    DVS27-related protein|IL1F11|IL-33
  • Protein/Peptide Name:
    IL33
  • Molecular Weight:
    19,2 kDa
  • Sequence:
    MKHHHHHHAS SITGISPITE YLASLSTYND QSITFALEDE SYEIYVEDLK KDEKKDKVLL SYYESQHPSN ESGDGVDGKM LMVTLSPTKD FWLHANNKEH SVELHKCEKP LPDQAFFVLH NMHSNCVSFE CKTDPGVFIG VKDNHLALIK VDSSENLCTE NILFKLSET
  • Cat. No.:
    102552-864
  • Supplier no.:
    RD172203100

Specifications

About this item

Total 169 AA. MW: 19,2 kDa (calculated). UniProtKB acc.no. O95760. N-Terminal His-tag, 10 extra AA

Sequence: MKHHHHHHAS SITGISPITE YLASLSTYND QSITFALEDE SYEIYVEDLK KDEKKDKVLL SYYESQHPSN ESGDGVDGKM LMVTLSPTKD FWLHANNKEH SVELHKCEKP LPDQAFFVLH NMHSNCVSFE CKTDPGVFIG VKDNHLALIK VDSSENLCTE NILFKLSET

Other names: IL-33, DVS27-related protein, IL1F11

Ordering information: For research use only.