Order Entry
Puerto Rico
ContactUsLinkComponent
Human Recombinant SFRP4 (from HEK293 Cells)
Human Recombinant SFRP4 (from HEK293 Cells)
Catalog # 101189-780
Supplier:  BioVendor
CAS Number:  
undefined
Human Recombinant SFRP4 (from HEK293 Cells)
Catalog # 101189-780
Supplier:  BioVendor
Supplier Number:  RD172132100
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Protein/Peptide Type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Size:
    100 µg
  • Protein/Peptide Name:
    SFRP4
  • Sequence:
    PGDYKDDDDK PAGVRGAPCE AVRIPMCRHM PWNITRMPNH LHHSTQENAI LAIEQYEELV DVNCSAVLRF FLCAMYAPIC TLEFLHDPIK PCKSVCQRAR DDCEPLMKMY NHSWPESLAC DELPVYDRGV CISPEAIVTD LPEDVKWIDI TPDMMVQERP LDVDCKRLSP DRCKCKKVKP TLATYLSKNY SYVIHAKIKA VQRSGCNEVT TVVDVKEIFK SSSPIPRTQV PLITNSSCQC PHILPHQDVL IMCYEWRSRM MLLENCLVEK WRDQLSKRSI QWEERLQEQR RTVQDKKKTA GRTSRSNPPK PKGKPPAPKP ASPKKNIKTR SAQKRTNPKR V
  • Tested Applications:
    Cell culture and/or animal studies, ELISA, Western Blot
  • Cat. No.:
    101189-780
  • Supplier no.:
    RD172132100

Specifications

About this item

sFRP-4 is a cystein-rich glycoprotein that belongs to the group of G protein-coupled receptor proteins. It is expressed in proliferative endometrium and several types of ovarian, endometrial and brest tumors. It is up-regulated in failing myocardium.

Sequence: PGDYKDDDDK PAGVRGAPCE AVRIPMCRHM PWNITRMPNH LHHSTQENAI LAIEQYEELV DVNCSAVLRF FLCAMYAPIC TLEFLHDPIK PCKSVCQRAR DDCEPLMKMY NHSWPESLAC DELPVYDRGV CISPEAIVTD LPEDVKWIDI TPDMMVQERP LDVDCKRLSP DRCKCKKVKP TLATYLSKNY SYVIHAKIKA VQRSGCNEVT TVVDVKEIFK SSSPIPRTQV PLITNSSCQC PHILPHQDVL IMCYEWRSRM MLLENCLVEK WRDQLSKRSI QWEERLQEQR RTVQDKKKTA GRTSRSNPPK PKGKPPAPKP ASPKKNIKTR SAQKRTNPKR V

Other names: SFRP4

Ordering information: For research use only.