Order Entry
Puerto Rico
ContactUsLinkComponent
Phosphoethanolamine/phosphocholine phosphatase Human E. coli, 0.1 mg
Phosphoethanolamine/phosphocholine phosphatase Human E. coli, 0.1 mg
Catalog # 101422-270
Supplier:  BioVendor
CAS Number:  
undefined
Phosphoethanolamine/phosphocholine phosphatase Human E. coli, 0.1 mg
Catalog # 101422-270
Supplier:  BioVendor
Supplier Number:  RD172117100
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Conjugation:
    Unconjugated
  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Size:
    0.1 mg
  • Storage Conditions:
    Store the lyophilized protein at –80 °C. Lyophilized protein remains stable until the expiry date when stored at –80 °C. Aliquot reconstituted protein to avoid repeated freezing/thawing cycles and store at –80 °C for long term storage. Reconstituted protein can be stored at 4 °C for three days.
  • Endotoxin Content:
    <0.1 EU/μg
  • Reconstitution Instructions:
    Add deionized water to prepare a working stock solution of approximately 0.25 mg/ml and let the lyophilized pellet dissolve completely.
  • Protein Synonyms:
    EC=3.1.3.75|PHOSPHO1|PHOSPHO-1
  • Protein/Peptide Name:
    Phosphoethanolamine/phosphocholine phosphatase Human E. coli
  • Purity:
    >95%
  • Molecular Weight:
    31.3 kDa (calculated)
  • Sequence:
    MRGSHHHHHHGMASMSGCFPVSGLRCLSRDGRMAAQGAPRFLLTFDFDETIVDENSDDSIVRAAPGQRLPESLRATYREGFYNEYMQRVFKYLGEQGVRPRDLSAIYEAIPLSPGMSDLLQFVAKQGACFEVILISDANTFGVESSLRAAGHHSLFRRILSNPSGPDARGLLALRPFHTHSCARCPANMCKHKVLSDYLRERAHDGVHFERLFYVGDGANDFCPMGLLAGGDVAFPRRGYPMHRLIQEAQKAEPSSFRASVVPWETAADVRLHLQQVLKSC
  • Endotoxin Level:
    <0.1 EU/μg
  • Formulation:
    Filtered (0.4 μm) and lyophilized from 0.5 mg/ml solution in phosphate buffered saline pH 7.5 + 5%(w/v) trehalose
  • Shipping Temperature:
    At ambient temperature
  • Tested Applications:
    Western blotting
  • Cat. No.:
    101422-270
  • Supplier no.:
    RD172117100

Specifications

About this item

Total 295 AA. MW: 31.3 kDa (calculated). UniProtKB acc.no. Q8TCT1. N-Terminal His-tag 14 AA (highlighted).

  • Quality control test
  • BCA to determine quantity of the protein
  • SDS PAGE to determine purity of the protein
  • LAL to determine quantity of endotoxin
Caution: Research use only.